Assay Details
Target Gene Details
Entrez Gene ID: | 11276 |
Gene Name: | synergin, gamma |
Gene Aliases: |
AP1GBP1, SYNG |
Location: |
Chr.17:37514797-37613637 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3640494 | Chr.17:37406555 - 37610947 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map