Assay Details
Target Gene Details
Entrez Gene ID: | 5596 |
Gene Name: | mitogen-activated protein kinase 4 |
Gene Aliases: |
ERK-4, ERK4, PRKM4, p63-MAPK, p63MAPK |
Location: |
Chr.18:50559781-50731826 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK4 | NM_001292039.1 | NP_001278968.1 | ||
| NM_001292040.1 | NP_001278969.1 | |||
| NM_002747.3 | NP_002738.2 | |||
| XM_011526076.2 | XP_011524378.1 | |||
| XM_017025839.1 | XP_016881328.1 | |||
| AK293818.1 | ||||
| AK295058.1 | ||||
| BC050299.1 | ||||
| BX648603.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34028 | Chr.18:50358141 - 50744133 on Build GRCh38 | Loss |
|
| nsv9630 | Chr.18:50532107 - 50922466 on Build GRCh38 | Loss |
|
| nsv833651 | Chr.18:50568263 - 50712984 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117221153] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map