Assay Details
Target Gene Details
Entrez Gene ID: | 6925 |
Gene Name: | transcription factor 4 |
Gene Aliases: |
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19 |
Location: |
Chr.18:55222331-55635993 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCF4 | NM_001083962.1 | 20 | 7597 | NP_001077431.1 |
| NM_001243226.2 | 21 | 7387 | NP_001230155.2 | |
| NM_001243227.1 | 19 | 7431 | NP_001230156.1 | |
| NM_001243228.1 | 20 | 7541 | NP_001230157.1 | |
| NM_001243230.1 | 19 | 7058 | NP_001230159.1 | |
| NM_001243231.1 | 18 | 7019 | NP_001230160.1 | |
| NM_001243232.1 | 16 | 7053 | NP_001230161.1 | |
| NM_001243233.1 | 16 | 6982 | NP_001230162.1 | |
| NM_001243234.1 | 13 | 6626 | NP_001230163.1 | |
| NM_001243235.1 | 13 | 6614 | NP_001230164.1 | |
| NM_001243236.1 | 13 | 6630 | NP_001230165.1 | |
| NM_001306207.1 | 19 | 7210 | NP_001293136.1 | |
| NM_001306208.1 | 16 | 7041 | NP_001293137.1 | |
| NM_003199.2 | 20 | 7585 | NP_003190.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2315 | Chr.18:55179705 - 55224322 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map