Assay Details
Target Gene Details
Entrez Gene ID: | 6925 |
Gene Name: | transcription factor 4 |
Gene Aliases: |
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19 |
Location: |
Chr.18:55222331-55635993 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Target Gene Details
Entrez Gene ID: | 105372127 |
Gene Name: | TCF4 antisense RNA 1 |
Gene Aliases: |
MIR4529HG |
Location: |
Chr.18:55452533-55482940 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCF4-AS1 | NR_132985.1 | |||
| AI027479.1 | ||||
| AI151133.1 | ||||
| AI419430.1 | ||||
| AI422931.1 | ||||
| AW138360.1 | ||||
| BX110727.1 |
More Information
Set Membership: |
|

Genomic Map