Assay Details
Target Gene Details
Entrez Gene ID: | 101927323 |
Gene Name: | PPP4R1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.18:9614807-9619363 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP4R1-AS1 | NR_110773.1 | |||
| NR_110774.1 | ||||
| BX108181.1 | ||||
| HY048010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521175 | Chr.18:9615853 - 9623054 on Build GRCh38 | Loss |
|
| nsv576448 | Chr.18:9558242 - 9828451 on Build GRCh38 | Gain |
|

Genomic Map