Assay Details
Target Gene Details
Entrez Gene ID: | 753 |
Gene Name: | low density lipoprotein receptor class A domain containing 4 |
Gene Aliases: |
C18orf1 |
Location: |
Chr.18:13217691-13652754 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LDLRAD4 | NM_181481.4 | NP_852146.1 | ||
| NM_181482.4 | NP_852147.1 | |||
| XM_005258140.1 | XP_005258197.1 | |||
| XM_006722353.1 | XP_006722416.1 | |||
| XM_006722354.1 | XP_006722417.1 | |||
| XM_011525737.1 | XP_011524039.1 | |||
| XM_011525738.1 | XP_011524040.1 | |||
| XM_017025961.1 | XP_016881450.1 | |||
| XM_017025962.1 | XP_016881451.1 | |||
| XM_017025963.1 | XP_016881452.1 | |||
| XM_017025964.1 | XP_016881453.1 | |||
| XM_017025965.1 | XP_016881454.1 | |||
| XM_017025966.1 | XP_016881455.1 | |||
| XM_017025967.1 | XP_016881456.1 | |||
| XM_017025968.1 | XP_016881457.1 | |||
| AF009424.1 | AAC52023.1 | |||
| AF009425.1 | AAC52024.1 | |||
| BC030199.1 | AAH30199.1 |
Target Gene Details
Entrez Gene ID: | 100288122 |
Gene Name: | LDLRAD4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.18:13419499-13427534 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LDLRAD4-AS1 | NR_040031.1 | |||
| AK098298.1 | ||||
| DT217238.1 |
More Information
Set Membership: |
|

Genomic Map