Assay Details
Target Gene Details
Entrez Gene ID: | 100652770 |
Gene Name: | DSG2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.18:31542146-31556911 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSG2-AS1 | NR_045216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576635 | Chr.18:31549968 - 31568851 on Build GRCh38 | Gain |
|
| nsv833615 | Chr.18:31428433 - 31613013 on Build GRCh38 | Gain |
|

Genomic Map