Assay Details
Target Gene Details
Entrez Gene ID: | 7050 |
Gene Name: | TGFB induced factor homeobox 1 |
Gene Aliases: |
HPE4, TGIF |
Location: |
Chr.18:3411927-3458412 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGIF1 | NM_174886.2 | NP_777480.1 | ||
| BE906361.1 | ||||
| CB996365.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2190 | Chr.18:3408994 - 3442002 on Build GRCh38 | Insertion |
|
| esv3641597 | Chr.18:3389935 - 3421195 on Build GRCh38 | Gain |
|
| esv3641584 | Chr.18:3200019 - 3415247 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map