Assay Details
Target Gene Details
Entrez Gene ID: | 6925 |
Gene Name: | transcription factor 4 |
Gene Aliases: |
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19 |
Location: |
Chr.18:55222331-55635993 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
More Information
Additional Information:
For this assay, SNP(s) [rs146599516] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map