Assay Details
Target Gene Details
Entrez Gene ID: | 147495 |
Gene Name: | APC down-regulated 1 |
Gene Aliases: |
B7323, DRAPC1, FP7019, HHS, HTS, HYPT1 |
Location: |
Chr.18:10454628-10488701 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APCDD1 | NM_153000.4 | NP_694545.1 | ||
| XM_011525603.1 | XP_011523905.1 | |||
| AB056722.1 | BAC15563.1 | |||
| AB104887.1 | BAC65165.1 | |||
| AK300743.1 | ||||
| BC053324.1 | AAH53324.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3055 | Chr.18:10462379 - 10462808 on Build GRCh38 | Loss |
|
| esv2716786 | Chr.18:10461975 - 10463011 on Build GRCh38 | Deletion |
|
| esv5818 | Chr.18:10462328 - 10462799 on Build GRCh38 | Loss |
|
| esv2443287 | Chr.18:10461911 - 10463318 on Build GRCh38 | Deletion |
|
| nsv1059110 | Chr.18:10142986 - 10596015 on Build GRCh38 | Gain |
|
| esv1968192 | Chr.18:10462250 - 10462824 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map