Assay Details
Target Gene Details
Entrez Gene ID: | 100192426 |
Gene Name: | uncharacterized LOC100192426 |
Gene Aliases: |
- |
Location: |
Chr.18:8360820-8367034 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100192426 | NR_024419.1 | |||
| AI184987.1 | ||||
| BX115569.1 |
Target Gene Details
Entrez Gene ID: | 5797 |
Gene Name: | protein tyrosine phosphatase, receptor type M |
Gene Aliases: |
PTPRL1, R-PTP-MU, RPTPM, RPTPU, hR-PTPu |
Location: |
Chr.18:7567316-8406861 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058445 | Chr.18:8270683 - 8421251 on Build GRCh38 | Loss |
|
| nsv526300 | Chr.18:6980524 - 8391831 on Build GRCh38 | Loss |
|
| nsv576426 | Chr.18:8110807 - 8645909 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map