Assay Details
Target Gene Details
Entrez Gene ID: | 29919 |
Gene Name: | chromosome 18 open reading frame 8 |
Gene Aliases: |
HsT2591, MIC1, Mic-1 |
Location: |
Chr.18:23503470-23531807 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C18orf8 | NM_001276342.1 | NP_001263271.1 | ||
| NM_001318707.1 | NP_001305636.1 | |||
| NM_001318708.1 | NP_001305637.1 | |||
| NM_001318709.1 | NP_001305638.1 | |||
| NM_013326.4 | NP_037458.3 | |||
| XM_011525939.1 | XP_011524241.1 | |||
| XM_017025717.1 | XP_016881206.1 | |||
| AF143536.1 | AAD33909.1 | |||
| AK057192.1 | BAB71377.1 | |||
| AK294169.1 | ||||
| AK295246.1 | ||||
| AK296230.1 | ||||
| BC002950.1 | AAH02950.2 | |||
| BC008305.1 | AAH08305.2 | |||
| CN481504.1 | ||||
| DC383182.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34149 | Chr.18:23278175 - 23641927 on Build GRCh38 | Loss |
|
| nsv509688 | Chr.18:23483859 - 23610653 on Build GRCh38 | Insertion |
|

Genomic Map