Assay Details
Target Gene Details
Entrez Gene ID: | 25925 |
Gene Name: | zinc finger protein 521 |
Gene Aliases: |
EHZF, Evi3 |
Location: |
Chr.18:25061924-25352250 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF521 | NM_001308225.1 | NP_001295154.1 | ||
| NM_015461.2 | NP_056276.1 | |||
| XM_011525909.2 | XP_011524211.1 | |||
| XM_011525911.1 | XP_011524213.1 | |||
| XM_017025697.1 | XP_016881186.1 | |||
| XM_017025698.1 | XP_016881187.1 | |||
| AF141339.1 | AAG49442.1 | |||
| AJ518106.1 | CAD57322.1 | |||
| AK021452.1 | BAB13829.1 | |||
| AK027354.1 | BAB55056.1 | |||
| AK074046.1 | ||||
| AK293067.1 | ||||
| AL117615.1 | CAB56016.1 | |||
| BC032869.2 | AAH32869.2 | |||
| BC113622.1 | ||||
| BC113648.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76425582] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map