Assay Details
Target Gene Details
Entrez Gene ID: | 1005 |
Gene Name: | cadherin 7 |
Gene Aliases: |
CDH7L1 |
Location: |
Chr.18:65750252-65885136 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH7 | NM_001317214.1 | NP_001304143.1 | ||
| NM_004361.3 | NP_004352.2 | |||
| NM_033646.2 | NP_387450.1 | |||
| XM_017025523.1 | XP_016881012.1 | |||
| AB035301.1 | BAA87415.1 | |||
| AJ007611.1 | CAC13127.1 | |||
| BC036786.1 | AAH36786.1 | |||
| BM560514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1067311 | Chr.18:63818011 - 66637685 on Build GRCh38 | Loss |
|
| nsv833675 | Chr.18:65631834 - 65807471 on Build GRCh38 | Gain |
|
| nsv577157 | Chr.18:65715200 - 68002467 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77530609] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map