Assay Details
Target Gene Details
Entrez Gene ID: | 114799 |
Gene Name: | establishment of sister chromatid cohesion N-acetyltransferase 1 |
Gene Aliases: |
A930014I12Rik, CTF, ECO1, EFO1, ESO1 |
Location: |
Chr.18:21529281-21601015 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESCO1 | NM_052911.2 | NP_443143.2 | ||
| XM_011525798.1 | XP_011524100.1 | |||
| XM_011525799.2 | XP_011524101.1 | |||
| AB067498.1 | ||||
| AK090579.1 | BAC03483.1 | |||
| AL832041.1 | CAH10584.1 | |||
| AL834200.1 | CAH10682.1 | |||
| BC036943.1 | AAH36943.1 | |||
| BC089426.1 | AAH89426.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3641938 | Chr.18:21412340 - 21588139 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map