Assay Details
Target Gene Details
Entrez Gene ID: | 57534 |
Gene Name: | mindbomb E3 ubiquitin protein ligase 1 |
Gene Aliases: |
DIP-1, DIP1, LVNC7, MIB, ZZANK2, ZZZ6 |
Location: |
Chr.18:21740793-21870957 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIB1 | NM_020774.3 | NP_065825.1 | ||
| XM_011526098.1 | XP_011524400.1 | |||
| XM_017025873.1 | XP_016881362.1 | |||
| AK091266.1 | ||||
| AY147849.1 | AAN18023.1 | |||
| AY149908.1 | AAN75493.1 | |||
| BC022403.1 | AAH22403.1 | |||
| BC110581.1 | AAI10582.1 | |||
| BC110582.1 | AAI10583.1 | |||
| CR749635.1 | CAH18429.1 |
Target Gene Details
Entrez Gene ID: | 102723167 |
Gene Name: | MIR133A1 host gene |
Gene Aliases: |
MIR1-2HG |
Location: |
Chr.18:21825479-21831406 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR133A1HG | NR_110369.1 | 3 | 3653 | |
| AK123912.1 | 1 | 1195 | ||
| BX648566.1 | 3 | 3654 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2222 | Chr.18:21816550 - 21851554 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map