Assay Details
Target Gene Details
Entrez Gene ID: | 7050 |
Gene Name: | TGFB induced factor homeobox 1 |
Gene Aliases: |
HPE4, TGIF |
Location: |
Chr.18:3411927-3458412 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGIF1 | NM_001278686.1 | NP_001265615.1 | ||
| NM_174886.2 | NP_777480.1 | |||
| BE906361.1 | ||||
| BM849453.1 | ||||
| CB996365.1 |
More Information
Set Membership: |
|

Genomic Map