Assay Details
Target Gene Details
Entrez Gene ID: | 125488 |
Gene Name: | tetratricopeptide repeat domain 39C |
Gene Aliases: |
C18orf17, HsT2697 |
Location: |
Chr.18:23992773-24135952 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC39C | NM_153211.3 | NP_694943.2 | ||
| XM_017025550.1 | XP_016881039.1 | |||
| AK091080.1 | BAC03578.1 | |||
| BC121034.2 | ||||
| BC121035.2 |
Target Gene Details
Entrez Gene ID: | 102724246 |
Gene Name: | TTC39C antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.18:23994213-24015460 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC39C-AS1 | NR_110796.1 | |||
| DB513641.1 |

Genomic Map