Assay Details
Target Gene Details
Entrez Gene ID: | 10216 |
Gene Name: | proteoglycan 4 |
Gene Aliases: |
CACP, HAPO, JCAP, MSF, SZP |
Location: |
Chr.1:186296273-186314562 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRG4 | NM_001127708.2 | NP_001121180.2 | ||
| NM_001127709.2 | NP_001121181.2 | |||
| NM_001127710.2 | NP_001121182.2 | |||
| NM_001303232.1 | NP_001290161.1 | |||
| NM_005807.4 | NP_005798.3 | |||
| XM_017000002.1 | XP_016855491.1 | |||
| XM_017000003.1 | XP_016855492.1 | |||
| AK057197.1 | ||||
| AK131434.1 | BAD18580.1 | |||
| AK297718.1 | ||||
| AK301343.1 | ||||
| AK301487.1 | ||||
| AK308235.1 | ||||
| DA977744.1 | ||||
| DA995681.1 | ||||
| DC295855.1 | ||||
| DC404857.1 | ||||
| U70136.1 | AAB09089.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575361 | Chr.1:186266048 - 186604612 on Build GRCh38 | Gain |
|
| esv3890826 | Chr.1:185730915 - 186448352 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map