Assay Details
Target Gene Details
Entrez Gene ID: | 128178 |
Gene Name: | EDAR associated death domain |
Gene Aliases: |
ECTD11A, ECTD11B, ED3, EDA3 |
Location: |
Chr.1:236394380-236484708 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EDARADD | NM_080738.3 | NP_542776.1 | ||
| NM_145861.2 | NP_665860.2 | |||
| AK096339.1 | ||||
| AK290862.1 | ||||
| AK291930.1 | ||||
| AK314634.1 | ||||
| AY028913.1 | AAK40287.1 | |||
| AY028914.1 | AAK40288.1 | |||
| AY071862.1 | AAL60590.1 | |||
| BC128082.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005877 | Chr.1:235912335 - 237038508 on Build GRCh38 | Gain |
|
| esv2763410 | Chr.1:236334208 - 236735742 on Build GRCh38 | Gain |
|
| nsv820827 | Chr.1:236421051 - 236422287 on Build GRCh38 | Deletion |
|
| nsv549394 | Chr.1:235876577 - 236629706 on Build GRCh38 | Gain |
|
| nsv1012436 | Chr.1:235860657 - 236648100 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113999596] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map