Assay Details
Target Gene Details
Entrez Gene ID: | 26523 |
Gene Name: | argonaute 1, RISC catalytic component |
Gene Aliases: |
EIF2C, EIF2C1, GERP95, Q99, hAgo1 |
Location: |
Chr.1:35869808-35929610 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGO1 | XM_011541236.2 | 19 | 11244 | XP_011539538.1 |
| XM_017001016.1 | 19 | 11292 | XP_016856505.1 | |
| XM_017001017.1 | 19 | 11283 | XP_016856506.1 | |
| XM_017001018.1 | 16 | 10767 | XP_016856507.1 | |
| XM_017001019.1 | 19 | 11332 | XP_016856508.1 | |
| AK025726.1 | 1 | 1460 | ||
| AK125879.1 | 1 | 2254 |
Target Gene Details
Entrez Gene ID: | 192669 |
Gene Name: | argonaute 3, RISC catalytic component |
Gene Aliases: |
EIF2C3 |
Location: |
Chr.1:35925198-36072037 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGO3 | XM_005270575.3 | 1 | 2883 | XP_005270632.1 |
| XM_017000523.1 | 1 | 2883 | XP_016856012.1 |
More Information
Set Membership: |
|

Genomic Map