Assay Details
Target Gene Details
Entrez Gene ID: | 1718 |
Gene Name: | 24-dehydrocholesterol reductase |
Gene Aliases: |
DCE, Nbla03646, SELADIN1, seladin-1 |
Location: |
Chr.1:54849627-54887248 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHCR24 | NM_014762.3 | NP_055577.1 | ||
| AB073391.1 | BAE45721.1 | |||
| AF261758.1 | AAG17288.1 | |||
| AK298414.1 | ||||
| AK302774.1 | ||||
| AK304302.1 | ||||
| AK304421.1 | ||||
| AK304437.1 | ||||
| AK316419.1 | ||||
| AK316427.1 | ||||
| BC004375.1 | AAH04375.1 | |||
| BC011669.2 | AAH11669.1 | |||
| D13643.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461561 | Chr.1:54404143 - 54887697 on Build GRCh38 | Gain |
|
| nsv817614 | Chr.1:54580862 - 55735351 on Build GRCh38 | Gain |
|
| nsv829915 | Chr.1:54828940 - 54992690 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map