Assay Details
Target Gene Details
Entrez Gene ID: | 284677 |
Gene Name: | chromosome 1 open reading frame 204 |
Gene Aliases: |
- |
Location: |
Chr.1:159834474-159855347 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf204 | NM_001134233.1 | NP_001127705.1 | ||
| AK096506.1 |
Target Gene Details
Entrez Gene ID: | 391123 |
Gene Name: | V-set and immunoglobulin domain containing 8 |
Gene Aliases: |
- |
Location: |
Chr.1:159854316-159862657 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VSIG8 | NM_001013661.1 | NP_001013683.1 | ||
| BC132893.1 | ||||
| BC132895.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005195 | Chr.1:159482969 - 159940547 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map