Assay Details
Target Gene Details
Entrez Gene ID: | 11243 |
Gene Name: | polyamine-modulated factor 1 |
Gene Aliases: |
- |
Location: |
Chr.1:156212988-156240077 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PMF1 | NM_001199653.1 | NP_001186582.1 | ||
| NM_001199654.1 | NP_001186583.1 | |||
| NM_007221.3 | NP_009152.2 | |||
| AF141310.1 | AAD50081.1 | |||
| AK022523.1 | ||||
| AK124646.1 | BAC85916.1 | |||
| AK289490.1 | ||||
| AK290260.1 | ||||
| AL080101.1 | CAH10730.1 | |||
| BC050735.1 | AAH50735.1 | |||
| BC056417.1 | AAH56417.1 | |||
| BC065031.1 | AAH65031.1 | |||
| DT220098.1 |
Target Gene Details
Entrez Gene ID: | 100527963 |
Gene Name: | PMF1-BGLAP readthrough |
Gene Aliases: |
PMF1 |
Location: |
Chr.1:156212988-156243332 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PMF1-BGLAP | NM_001199661.1 | NP_001186590.1 | ||
| NM_001199662.1 | NP_001186591.1 | |||
| NM_001199663.1 | NP_001186592.1 | |||
| NM_001199664.1 | NP_001186593.1 | |||
| CD049192.1 | ||||
| CD388852.1 | ||||
| FY213424.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509524 | Chr.1:156238274 - 156308437 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map