Assay Details
Target Gene Details
Entrez Gene ID: | 4359 |
Gene Name: | myelin protein zero |
Gene Aliases: |
CHM, CMT1, CMT1B, CMT2I, CMT2J, CMT4E, CMTDI3, CMTDID, DSS, HMSNIB, MPP, P0 |
Location: |
Chr.1:161303593-161309972 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPZ | NM_000530.7 | NP_000521.2 | ||
| NM_001315491.1 | NP_001302420.1 | |||
| XM_017001321.1 | XP_016856810.1 | |||
| AK313555.1 | ||||
| BT006765.1 | AAP35411.1 | |||
| D10537.1 | BAA01395.1 | |||
| DB210759.1 | ||||
| S66705.1 | AAB28708.1 |
More Information
Set Membership: |
|

Genomic Map