Assay Details
Target Gene Details
Entrez Gene ID: | 6279 |
Gene Name: | S100 calcium binding protein A8 |
Gene Aliases: |
60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8 |
Location: |
Chr.1:153390032-153422583 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A8 | NM_001319198.1 | NP_001306127.1 | ||
| AA401538.1 | ||||
| AI126134.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002705 | Chr.1:153335376 - 153553113 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74115420,rs74876421] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map