Assay Details
Target Gene Details
Entrez Gene ID: | 6814 |
Gene Name: | syntaxin binding protein 3 |
Gene Aliases: |
MUNC18-3, MUNC18C, PSP, UNC-18C |
Location: |
Chr.1:108746663-108809526 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STXBP3 | NM_007269.2 | NP_009200.2 | ||
| AF032922.1 | AAC69606.1 | |||
| AK223170.1 | BAD96890.1 | |||
| AK290134.1 | ||||
| AK291322.1 | ||||
| AK312910.1 | ||||
| BC038099.2 | AAH38099.1 | |||
| BC047764.1 | AAH47764.1 | |||
| D63506.1 | BAA19482.1 |
More Information
Set Membership: |
|

Genomic Map