Assay Details
Target Gene Details
Entrez Gene ID: | 5784 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 14 |
Gene Aliases: |
PEZ, PTP36 |
Location: |
Chr.1:214348696-214551681 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPN14 | NM_005401.4 | 19 | 12395 | NP_005392.2 |
| AF086517.1 | 1 | 487 | ||
| AK056963.1 | 1 | 61 |
More Information
Set Membership: |
|

Genomic Map