Assay Details
Target Gene Details
Entrez Gene ID: | 100852410 |
Gene Name: | ZRANB2 antisense RNA 2 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.1:71081324-71237723 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZRANB2-AS2 | NR_046217.1 | |||
| BM766420.1 |

Genomic Map