Assay Details
Target Gene Details
Entrez Gene ID: | 84976 |
Gene Name: | dispatched RND transporter family member 1 |
Gene Aliases: |
DISPA |
Location: |
Chr.1:222814514-223005995 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DISP1 | NM_032890.3 | NP_116279.2 | ||
| XM_011510072.2 | XP_011508374.1 | |||
| XM_011510073.2 | XP_011508375.1 | |||
| XM_011510074.2 | XP_011508376.1 | |||
| XM_011510075.2 | XP_011508377.1 | |||
| XM_011510077.2 | XP_011508379.1 | |||
| XM_017002611.1 | XP_016858100.1 | |||
| DA756409.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv442589 | Chr.1:222843983 - 222850262 on Build GRCh38 | Loss |
|
| nsv1160101 | Chr.1:222843064 - 222850427 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117818119] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map