Assay Details
Target Gene Details
Entrez Gene ID: | 57489 |
Gene Name: | outer dense fiber of sperm tails 2 like |
Gene Aliases: |
dJ977L11.1 |
Location: |
Chr.1:86346824-86396345 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2714374 | Chr.1:86385812 - 86385985 on Build GRCh38 | Deletion |
|
| nsv546722 | Chr.1:85812632 - 86425390 on Build GRCh38 | Loss |
|
| esv2714363 | Chr.1:86385710 - 86386129 on Build GRCh38 | Deletion |
|
| nsv1002541 | Chr.1:86223405 - 86440219 on Build GRCh38 | Gain |
|
| esv2657167 | Chr.1:86385842 - 86385998 on Build GRCh38 | Deletion |
|

Genomic Map