Assay Details
Target Gene Details
Entrez Gene ID: | 391013 |
Gene Name: | phospholipase A2 group IIC |
Gene Aliases: |
- |
Location: |
Chr.1:20163085-20177364 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G2C | NM_001316722.1 | NP_001303651.1 | ||
| XM_017001270.1 | 1 | 118 | XP_016856759.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545731 | Chr.1:19989106 - 20776306 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map