Assay Details
Target Gene Details
Entrez Gene ID: | 80896 |
Gene Name: | N-acetylneuraminate pyruvate lyase |
Gene Aliases: |
C112, C1orf13, NAL, NPL1 |
Location: |
Chr.1:182789449-182830384 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPL | NM_001200050.1 | NP_001186979.1 | ||
| NM_001200051.1 | NP_001186980.1 | |||
| NM_001200052.1 | NP_001186981.1 | |||
| NM_001200056.1 | NP_001186985.1 | |||
| NM_030769.2 | NP_110396.1 | |||
| AF338436.1 | AAK25795.1 | |||
| AK297017.1 | ||||
| AK301068.1 | ||||
| AK313225.1 | ||||
| AL525867.3 | ||||
| AY336748.1 | AAQ82432.1 | |||
| BC058003.1 | AAH58003.1 | |||
| CB961482.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548358 | Chr.1:182110851 - 183151893 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map