Assay Details
Target Gene Details
Entrez Gene ID: | 6284 |
Gene Name: | S100 calcium binding protein A13 |
Gene Aliases: |
- |
Location: |
Chr.1:153618799-153634092 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A13 | NM_001024210.1 | NP_001019381.1 | ||
| XM_011509862.2 | XP_011508164.1 | |||
| XM_011509863.2 | XP_011508165.1 | |||
| XM_017002034.1 | XP_016857523.1 | |||
| XM_017002035.1 | XP_016857524.1 | |||
| CK817939.1 |
More Information
Set Membership: |
|

Genomic Map