Assay Details
Target Gene Details
Entrez Gene ID: | 919 |
Gene Name: | CD247 molecule |
Gene Aliases: |
CD3-ZETA, CD3H, CD3Q, CD3Z, IMD25, T3Z, TCRZ |
Location: |
Chr.1:167430640-167518610 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD247 | NM_000734.3 | NP_000725.1 | ||
| NM_198053.2 | NP_932170.1 | |||
| XM_011510144.2 | XP_011508446.1 | |||
| XM_011510145.1 | XP_011508447.1 | |||
| XM_017002800.1 | XP_016858289.1 | |||
| XM_017002801.1 | XP_016858290.1 | |||
| AF228312.1 | AAF34793.1 | |||
| AK128376.1 | 5 | 1353 | BAC87407.1 | |
| AK223325.1 | BAD97045.1 | |||
| AK313946.1 | ||||
| AL557555.3 | ||||
| BC025703.1 | AAH25703.1 | |||
| BI819937.1 | ||||
| EU117371.1 | ||||
| EU117372.1 | ||||
| EU117373.1 | ||||
| EU118788.1 | ||||
| EU118789.1 | ||||
| EU118790.1 | ||||
| EU118791.1 | ||||
| EU118792.1 | ||||
| EU118793.1 | ||||
| EU597245.1 | ||||
| J04132.1 | AAA60394.1 | |||
| X55510.1 | ||||
| X55511.1 | ||||
| X55512.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890749 | Chr.1:167379314 - 167446554 on Build GRCh38 | Gain |
|
| nsv3432 | Chr.1:167418866 - 167452640 on Build GRCh38 | Insertion |
|
| nsv831814 | Chr.1:167380597 - 167590974 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map