Assay Details
Target Gene Details
Entrez Gene ID: | 9214 |
Gene Name: | Fc fragment of IgM receptor |
Gene Aliases: |
FAIM3, TOSO |
Location: |
Chr.1:206903282-206922033 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCMR | NM_001142473.1 | NP_001135945.1 | ||
| NM_001193338.1 | NP_001180267.1 | |||
| NM_005449.4 | NP_005440.1 | |||
| XM_005273351.3 | XP_005273408.1 | |||
| XM_005273352.4 | XP_005273409.1 | |||
| AF057557.1 | AAC18830.1 | |||
| AK292007.1 | ||||
| AK297053.1 | ||||
| AK301187.1 | ||||
| AK316336.1 | ||||
| BC006401.2 | AAH06401.1 | |||
| BP225124.1 | ||||
| BP305798.1 | ||||
| BT006797.1 | AAP35443.1 | |||
| DA971987.1 | ||||
| DB122623.1 | ||||
| DC393681.1 | ||||
| GQ160900.1 | ||||
| GQ160901.1 | ||||
| HM480394.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010486 | Chr.1:206876486 - 207031685 on Build GRCh38 | Gain |
|
| nsv468049 | Chr.1:206894495 - 206963993 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115727949] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map