Assay Details
Target Gene Details
Entrez Gene ID: | 127733 |
Gene Name: | UBX domain protein 10 |
Gene Aliases: |
UBXD3 |
Location: |
Chr.1:20183106-20196060 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBXN10 | XM_011540699.2 | XP_011539001.1 |
Target Gene Details
Entrez Gene ID: | 101928017 |
Gene Name: | UBXN10 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:20184242-20186486 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBXN10-AS1 | NR_110078.1 | |||
| AA683341.1 | ||||
| AI651649.1 | ||||
| BX106299.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545731 | Chr.1:19989106 - 20776306 on Build GRCh38 | Gain |
|

Genomic Map