Assay Details
Target Gene Details
Entrez Gene ID: | 84251 |
Gene Name: | SH3 domain GRB2 like endophilin interacting protein 1 |
Gene Aliases: |
- |
Location: |
Chr.1:66533272-66751139 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
More Information
Set Membership: |
|

Genomic Map