Assay Details
Target Gene Details
Entrez Gene ID: | 10560 |
Gene Name: | solute carrier family 19 member 2 |
Gene Aliases: |
TC1, THMD1, THT1, THTR1, TRMA |
Location: |
Chr.1:169463909-169485970 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC19A2 | NM_001319667.1 | NP_001306596.1 | ||
| NM_006996.2 | NP_008927.1 | |||
| AB209540.1 | 1 | 119 | BAD92777.1 | |
| AF135488.1 | AAD45985.1 | |||
| AF153330.1 | AAD43534.1 | |||
| AF160812.1 | AAF15129.1 | |||
| AF272359.1 | AAK54468.1 | |||
| AJ237724.1 | CAB50770.1 | |||
| AK304021.1 | ||||
| AK313779.1 | ||||
| AK316465.1 | ||||
| BC018514.1 | AAH18514.1 | |||
| KU178534.1 | ||||
| KU178535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005941 | Chr.1:169441248 - 169487155 on Build GRCh38 | Loss |
|
| esv22143 | Chr.1:169090390 - 169652030 on Build GRCh38 | Gain+Loss |
|
| nsv1001454 | Chr.1:169418143 - 169496036 on Build GRCh38 | Gain |
|
| nsv831871 | Chr.1:169351435 - 169517442 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map