Assay Details
Target Gene Details
Entrez Gene ID: | 4881 |
Gene Name: | natriuretic peptide receptor 1 |
Gene Aliases: |
ANPRA, ANPa, GUC2A, GUCY2A, NPRA |
Location: |
Chr.1:153678617-153693992 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPR1 | NM_000906.3 | NP_000897.3 | ||
| XM_005245218.1 | XP_005245275.1 | |||
| AK025024.1 | ||||
| AK298090.1 | ||||
| AK300446.1 | ||||
| BC063304.1 | AAH63304.1 | |||
| S72628.1 | AAD14112.1 | |||
| X15357.1 | CAA33417.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464028 | Chr.1:153649744 - 153750561 on Build GRCh38 | Gain |
|
| dgv582n54 | Chr.1:153639657 - 153750561 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map