Assay Details
Target Gene Details
Entrez Gene ID: | 54896 |
Gene Name: | PQ loop repeat containing 2 |
Gene Aliases: |
- |
Location: |
Chr.1:19312246-19332611 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PQLC2 | NR_109848.1 | |||
| XM_005245916.2 | 8 | 3193 | XP_005245973.1 | |
| XM_006710710.2 | 9 | 3303 | XP_006710773.1 | |
| XM_006710711.2 | 8 | 3162 | XP_006710774.1 | |
| XM_011541634.2 | 8 | 4177 | XP_011539936.1 | |
| AK000327.1 | BAA91088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv21e212 | Chr.1:19324106 - 19361938 on Build GRCh38 | Loss |
|
| esv2674133 | Chr.1:19330124 - 19334699 on Build GRCh38 | Deletion |
|
| esv3585396 | Chr.1:19330138 - 19334718 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map