Assay Details
Target Gene Details
Entrez Gene ID: | 6464 |
Gene Name: | SHC adaptor protein 1 |
Gene Aliases: |
SHC, SHCA |
Location: |
Chr.1:154962298-154974492 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHC1 | NM_001130041.1 | NP_001123513.1 | ||
| NM_001202859.1 | NP_001189788.1 | |||
| NM_003029.4 | NP_003020.2 | |||
| XM_005245449.4 | 1 | 393 | XP_005245506.1 | |
| XM_005245451.4 | XP_005245508.1 | |||
| XM_011509892.2 | 1 | 390 | XP_011508194.1 | |
| XM_011509893.2 | 1 | 391 | XP_011508195.1 | |
| XM_011509894.2 | 1 | 394 | XP_011508196.1 | |
| XM_011509897.1 | XP_011508199.1 | |||
| XM_011509898.2 | XP_011508200.1 | |||
| XM_017002081.1 | 1 | 389 | XP_016857570.1 | |
| XM_017002082.1 | 1 | 390 | XP_016857571.1 | |
| XM_017002083.1 | XP_016857572.1 | |||
| AK130014.1 | ||||
| AK292143.1 | ||||
| AK315842.1 | ||||
| BC014158.2 | AAH14158.1 | |||
| X68148.1 | CAA48251.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map