Assay Details
Target Gene Details
Entrez Gene ID: | 55220 |
Gene Name: | kelch domain containing 8A |
Gene Aliases: |
- |
Location: |
Chr.1:205336065-205357090 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHDC8A | NM_001271863.1 | NP_001258792.1 | ||
| NM_001271864.1 | NP_001258793.1 | |||
| NM_001271865.1 | NP_001258794.1 | |||
| NM_018203.2 | NP_060673.1 | |||
| AK001610.1 | BAA91787.1 | |||
| AK096590.1 | ||||
| AK299227.1 | ||||
| BC036076.1 | AAH36076.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832381 | Chr.1:205290241 - 205479588 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map