Assay Details
Target Gene Details
Entrez Gene ID: | 27 |
Gene Name: | ABL proto-oncogene 2, non-receptor tyrosine kinase |
Gene Aliases: |
ABLL, ARG |
Location: |
Chr.1:179099327-179229693 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABL2 | NM_001136000.2 | 13 | 8499 | NP_001129472.1 |
| NM_001168236.1 | 11 | 9032 | NP_001161708.1 | |
| NM_001168237.1 | 13 | 8786 | NP_001161709.1 | |
| NM_001168238.1 | 12 | 8723 | NP_001161710.1 | |
| NM_001168239.1 | 12 | 8436 | NP_001161711.1 | |
| NM_005158.4 | 12 | 8808 | NP_005149.4 | |
| NM_007314.3 | 12 | 9095 | NP_009298.1 | |
| AK025877.1 | 1 | 1389 | ||
| AK056280.1 | 1 | 2268 |
More Information
Additional Information:
For this assay, SNP(s) [rs535394578] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map