Assay Details
Target Gene Details
Entrez Gene ID: | 5738 |
Gene Name: | prostaglandin F2 receptor inhibitor |
Gene Aliases: |
CD315, CD9P-1, EWI-F, FPRP, SMAP-6 |
Location: |
Chr.1:116909923-116990358 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGFRN | NM_020440.3 | NP_065173.2 | ||
| AB037857.1 | ||||
| AK074637.1 | BAC11104.1 | |||
| BC098142.1 | AAH98142.1 | |||
| BC114520.1 | AAI14521.1 | |||
| BC139718.1 | ||||
| BC152454.1 | ||||
| DA572787.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831159 | Chr.1:116844705 - 117040990 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73012746] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map