Assay Details
Target Gene Details
Entrez Gene ID: | 374973 |
Gene Name: | testis expressed 38 |
Gene Aliases: |
ATPAF1-AS1, C1orf223, THEG4 |
Location: |
Chr.1:46669344-46673603 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TEX38 | XM_011541421.2 | XP_011539723.1 |
More Information
Set Membership: |
|

Genomic Map