Assay Details
Target Gene Details
Entrez Gene ID: | 5563 |
Gene Name: | protein kinase AMP-activated catalytic subunit alpha 2 |
Gene Aliases: |
AMPK, AMPK2, AMPKa2, PRKAA |
Location: |
Chr.1:56645317-56715335 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAA2 | NM_006252.3 | NP_006243.2 | ||
| XM_017001692.1 | XP_016857181.1 | |||
| XM_017001693.1 | XP_016857182.1 | |||
| XM_017001694.1 | XP_016857183.1 | |||
| AK314011.1 | ||||
| BC069680.1 | AAH69680.1 | |||
| BC069740.1 | AAH69740.1 | |||
| BC069823.1 | AAH69823.1 | |||
| DA710012.1 | ||||
| EF056019.1 | ||||
| FJ200485.1 | ||||
| FJ200486.1 | ||||
| HQ628625.1 | ||||
| U06454.1 | AAA64745.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523032 | Chr.1:56651236 - 56783885 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map