Assay Details
Target Gene Details
Entrez Gene ID: | 3604 |
Gene Name: | TNF receptor superfamily member 9 |
Gene Aliases: |
4-1BB, CD137, CDw137, ILA |
Location: |
Chr.1:7915871-7943165 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFRSF9 | NM_001561.5 | NP_001552.2 | ||
| XM_006710618.3 | 1 | 51 | XP_006710681.1 | |
| BP381234.1 |
More Information
Set Membership: |
|

Genomic Map