Assay Details
Target Gene Details
Entrez Gene ID: | 2048 |
Gene Name: | EPH receptor B2 |
Gene Aliases: |
CAPB, DRT, EK5, EPHT3, ERK, Hek5, PCBC, Tyro5 |
Location: |
Chr.1:22710770-22921500 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHB2 | NM_001309192.1 | NP_001296121.1 | ||
| NM_001309193.1 | NP_001296122.1 | |||
| NM_004442.7 | NP_004433.2 | |||
| NM_017449.4 | NP_059145.2 | |||
| XM_006710442.3 | XP_006710505.1 | |||
| AB210018.1 | ||||
| AF025304.1 | AAB94602.1 | |||
| AK299701.1 | ||||
| BC067861.1 | AAH67861.1 | |||
| DB282024.1 | ||||
| L36643.1 | AAA74244.1 | |||
| L41939.1 | AAA99310.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34161 | Chr.1:22599762 - 22817730 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72879171] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map