Assay Details
Target Gene Details
Entrez Gene ID: | 10768 |
Gene Name: | adenosylhomocysteinase like 1 |
Gene Aliases: |
DCAL, IRBIT, PPP1R78, PRO0233, XPVKONA |
Location: |
Chr.1:109984741-110023742 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AHCYL1 | NM_001242673.1 | NP_001229602.1 | ||
| NM_001242674.1 | NP_001229603.1 | |||
| NM_006621.5 | NP_006612.2 | |||
| XM_011540535.1 | XP_011538837.1 | |||
| AF315687.1 | AAL26869.1 | |||
| AK303690.1 | ||||
| AK308080.1 | ||||
| BC007576.1 | ||||
| BC010681.1 | ||||
| BC016942.1 | ||||
| BC065254.1 | ||||
| BC095476.1 | ||||
| BC110896.1 | ||||
| BC111563.1 | AAI11564.1 | |||
| DC329490.1 | ||||
| U82761.1 | AAC01960.1 |
More Information
Set Membership: |
|

Genomic Map